Merck
  • Solution and in silico studies on the recombinant lectin from Cicer arietinum seeds.
Solution and in silico studies on the recombinant lectin from Cicer arietinum seeds.
International journal of biological macromolecules (2013-03-15)
Madhurima S Wakankar, Musti V Krishnasastry, Tulika M Jaokar, Krunal A Patel, Sushama M Gaikwad
ABSTRACT
The Cicer arietinum seed lectin was cloned and expressed in Escherichia coli and purified in active form. Conformational characterization of the recombinant lectin (rCAL) was performed using biophysical and bioinformatics tools. Thermal denaturation of rCAL caused rapid secondary structural rearrangements above 50 °C and transient exposure of hydrophobic residues at 55 °C, leading to aggregation. Treatment of rCAL with GdnHCl resulted in unfolding followed by dissociation of the dimer. The single tryptophan in rCAL present on the surface of the protein is surrounded by hydrophobic and acidic amino acids and exists as different conformers. The experimental observations correlated well with the structural information revealed from the homology model of rCAL.
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%