Merck
  • Bioinspired construction of a spirocyclohexadienone moiety via sodium nitrite catalyzed aerobic intramolecular oxidative phenol coupling.
Bioinspired construction of a spirocyclohexadienone moiety via sodium nitrite catalyzed aerobic intramolecular oxidative phenol coupling.
Organic letters (2013-03-13)
Bo Su, Meng Deng, Qingmin Wang
ABSTRACT
An efficient and green intramolecular oxidative phenol coupling for the direct construction of spirocyclohexadienones has been developed, which uses environment-friendly sodium nitrite as the catalyst and oxygen in the air as the terminal oxidant. Hydroxy-containing substituted phenanthrenes and dibenzoazepines could be easily obtained from the dienone-phenol rearrangement.
Product Description
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%