Merck
  • Serum cation profile of broilers at various stages of exposure to deoxynivalenol.
Serum cation profile of broilers at various stages of exposure to deoxynivalenol.
Mycotoxin research (2013-02-23)
Agha Waqar Yunus, Josef Böhm
ABSTRACT
The present experiment was carried out to investigate if levels of serum cations in broilers are modulated differently at various stages of exposure to deoxynivalenol (DON). Male broiler chicks at 7 days of age were fed a basal diet (0.27 mg of DON; 0.01 mg of zearalenone/kg), or either a low DON diet (1.68 mg of DON; 0.15 mg of zearalenone/kg) or a high DON diet (12.21 mg of DON; 1.09 mg of zearalenone/kg) produced using extracts from Fusarium graminearum cultures. Blood samples from the birds were collected during weeks 2, 4, and 5 of exposure. The high DON diet resulted in lower serum calcium levels compared to the basal diet at all the 3 sampling stages, while the low DON diet resulted in lower serum calcium levels only during weeks 2 and 5. Serum potassium levels were reduced under both the DON diets during weeks 2 and 5, while no diet-associated changes were found for serum levels of magnesium, sodium, and zinc. Under the present experimental conditions, the serum levels of calcium were consistently modulated in the broilers exposed to the DON-contaminated diets. The modulation of serum levels of potassium was, however, dependent upon the stage of exposure to DON.
Product Description
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Deoxynivalenol