Merck
  • 4-[18F]Fluoro-N-methyl-N-(propyl-2-yn-1-yl)benzenesulfonamide ([18F]F-SA): a versatile building block for labeling of peptides, proteins and oligonucleotides with fluorine-18 via Cu(I)-mediated click chemistry.
4-[18F]Fluoro-N-methyl-N-(propyl-2-yn-1-yl)benzenesulfonamide ([18F]F-SA): a versatile building block for labeling of peptides, proteins and oligonucleotides with fluorine-18 via Cu(I)-mediated click chemistry.
Amino acids (2013-01-12)
Theres Ramenda, Jörg Steinbach, Frank Wuest
ABSTRACT
Cu(I)-mediated [3+2]cycloaddition between azides and alkynes has evolved into a valuable bioconjugation tool in radiopharmaceutical chemistry. We have developed a simple, convenient and reliable radiosynthesis of 4-[18F]fluoro-N-methyl-N-(propyl-2-yn-1-yl)benzenesulfonamide ([18F]F-SA) as a novel aromatic sulfonamide-based click chemistry building block. [18F]F-SA could be prepared in a remotely controlled synthesis unit in 32 ± 5% decay-corrected radiochemical yield in a total synthesis time of 80 min. The determined lipophilicity of [18F]F-SA (logP = 1.7) allows handling of the radiotracer in aqueous solutions. The versatility of [18F]F-SA as click chemistry building block was demonstrated by the labeling of a model peptide (phosphopeptide), protein (HSA), and oligonucleotide (L-RNA). The obtained radiochemical yields were 77% (phosphopeptide), 55-60% (HSA), and 25% (L-RNA), respectively. Despite the recent emergence of a multitude of highly innovative novel bioconjugation methods for 18F labeling of biopolymers, Cu(I)-mediated click chemistry with [18F]F-SA represents a reliable, robust and efficient radiolabeling technique for peptides, proteins, and oligonucleotides with the short-lived positron emitter 18F.
Sigma-Aldrich
Benzenesulfonamide, ≥98%
Pricing and availability is not currently available.