Merck
  • Exploring the role of shelf sediments in the Arctic Ocean in determining the Arctic contamination potential of neutral organic contaminants.
Exploring the role of shelf sediments in the Arctic Ocean in determining the Arctic contamination potential of neutral organic contaminants.
Environmental science & technology (2012-12-18)
James M Armitage, Sung-Deuk Choi, Torsten Meyer, Trevor N Brown, Frank Wania
ABSTRACT
The main objective of this study was to model the contribution of shelf sediments in the Arctic Ocean to the total mass of neutral organic contaminants accumulated in the Arctic environment using a standardized emission scenario for sets of hypothetical chemicals and realistic emission estimates (1930-2100) for polychlorinated biphenyl congener 153 (PCB-153). Shelf sediments in the Arctic Ocean are shown to be important reservoirs for neutral organic chemicals across a wide range of partitioning properties, increasing the total mass in the surface compartments of the Arctic environment by up to 3.5-fold compared to simulations excluding this compartment. The relative change in total mass for hydrophobic organic chemicals with log air-water partition coefficients ≥0 was greater than for chemicals with properties similar to typical POPs. The long-term simulation of PCB-153 generated modeled concentrations in shelf sediments in reasonable agreement with available monitoring data and illustrate that the relative importance of shelf sediments in the Arctic Ocean for influencing surface ocean concentrations (and therefore exposure via the pelagic food web) is most pronounced once primary emissions are exhausted and secondary sources dominate. Additional monitoring and modeling work to better characterize the role of shelf sediments for contaminant fate is recommended.
Product Description
Supelco
PCB No 153, analytical standard
Pricing and availability is not currently available.
2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153), BCR®, certified reference material
Pricing and availability is not currently available.