Merck
  • Photochemical trans-/cis-isomerization and quantitation of zearalenone in edible oils.
Photochemical trans-/cis-isomerization and quantitation of zearalenone in edible oils.
Journal of agricultural and food chemistry (2012-11-20)
Robert Köppen, Juliane Riedel, Matthias Proske, Sarah Drzymala, Tatjana Rasenko, Vedat Durmaz, Marcus Weber, Matthias Koch
ABSTRACT
The emphasis of the present work was to investigate the photochemical conversion of trans- to cis-zearalenone in edible oils under real-life conditions. For quantitation purposes a cis-zearalenone standard was synthesized and characterized for its identity and purity (≥95%) by (1)H NMR, X-ray crystallography, HPLC fluorescence and mass spectrometric detection. In a sample survey of 12 edible oils (9 corn oils, 3 hempseed oils) from local supermarkets all corn oils contained trans-zearalenone (median 194 μg/kg), but no cis-zearalenone was detected. For alteration studies trans-zearalenone contaminated corn oils were exposed to sunlight over 4 and 30 weeks, revealing an obvious shift toward cis-zearalenone up to a cis/trans ratio of 9:1 by storage in colorless glass bottles. Irradiation experiments of trans-zearalenone in different organic solvents confirmed the preferred formation of cis-zearalenone possibly caused by entropic effects rather than by enthalpic entities as investigated by quantum chemical and classical force field simulations.