Merck
  • Madecassoside, a triterpenoid saponin isolated from Centella asiatica herbs, protects endothelial cells against oxidative stress.
Madecassoside, a triterpenoid saponin isolated from Centella asiatica herbs, protects endothelial cells against oxidative stress.
Journal of biochemical and molecular toxicology (2012-07-26)
Difei Bian, Mei Liu, Ying Li, Yufeng Xia, Zhunan Gong, Yue Dai
ABSTRACT
This study aimed to investigate the effect of madecassoside against oxidative stress-induced injury of endothelial cells. Hydrogen peroxide (H(2)O(2), 500 µmol/L) was employed as an inducer of oxidative stress in human umbilical vein endothelial cells (HUVECs). Cell apoptosis was detected by Hoechst 33258 staining and flow cytometry. Caspase-3 activity and mitochondria membrane potential were further examined. As a result, madecassoside (10, 30, 100 µmol/L) could reverse morphological changes, elevate cell viability, increase glutathione levels, and decrease lactate dehydrogenase and malondialdehyde levels caused by H(2)O(2) in a concentration-dependent manner. It attenuated apoptosis, preventing the activation of caspase-3 and the loss of mitochondria membrane potential, as well as the phosphorylation of p38 mitogen-activated protein kinase (MAPK) in HUVECs. These data suggested that madecassoside could protect HUVECs from oxidative injury, which was probably achieved by inhibiting cell apoptosis via protection of mitochondria membranes and downregulation of the activation of caspase-3 and p38 MAPK.
Sigma-Aldrich
Madecassoside from Centella asiatica, ≥95% (HPLC)