- Chemical probes for the recognition of cannabinoid receptors in native systems.
Chemical probes for the recognition of cannabinoid receptors in native systems.
Angewandte Chemie (International ed. in English) (2012-06-13)
Lidia Martín-Couce, Mar Martín-Fontecha, Oscar Palomares, Leyre Mestre, Arnau Cordomí, Miriam Hernangomez, Sara Palma, Leonardo Pardo, Carmen Guaza, María L López-Rodríguez, Silvia Ortega-Gutiérrez
PMID22689411
ABSTRACT
Receptors made visible: The described biotin-tagged small-molecule probes with excellent affinities for the CB(1) and CB(2) cannabinoid receptors (CB(1)R and CB(2)R) enable direct visualization of these receptors in native cellular systems, including neurons, microglia, and immune cells. This method could overcome some of the limitations of current methodologies and may help to dissect the complexity of the endogenous cannabinoid system.
Product Description
Supelco
HU-210 solution, 100 μg/mL in methanol, (Spice Cannabinoid), ampule of 1 mL, certified reference material, Cerilliant®