Merck
  • Evaluation of the interaction between β-cyclodextrin and psychotropic drugs by surface plasmon resonance assay with a Biacore ® system.
Evaluation of the interaction between β-cyclodextrin and psychotropic drugs by surface plasmon resonance assay with a Biacore ® system.
Journal of pharmaceutical and biomedical analysis (2010-09-15)
Hiroaki Kobayashi, Tomohiro Endo, Noriko Ogawa, Hiromasa Nagase, Mariko Iwata, Haruhisa Ueda
ABSTRACT
Phase-solubility studies have been used to evaluate the solubilizing effects of cyclodextrins (CDs) on lipophilic, water-insoluble drugs. However, large amounts of CDs and drugs are required to measure solubility by phase-solubility studies. Thus, more efficient approaches to evaluate the interaction of CDs with drugs are needed. Herein we introduce a method that evaluates the interaction between immobilized β-cyclodextrin and psychotropic drugs by surface plasmon resonance assay with a Biacore(®) system. Association constants and stoichiometries observed were generally consistent with values calculated by traditional methods, such as phase-solubility and continuous variation methods. Results showed that the analytical method using Biacore(®) was suitable to evaluate CD-drug interactions.