- Long-term trends of polychlorinated biphenyls and chlorinated pesticides in franciscana dolphin (Pontoporia blainvillei) from Southern Brazil.
Long-term trends of polychlorinated biphenyls and chlorinated pesticides in franciscana dolphin (Pontoporia blainvillei) from Southern Brazil.
Marine pollution bulletin (2009-11-26)
Juliana Leonel, Jose L Sericano, Gilberto Fillmann, Eduardo Secchi, Rosalinda C Montone
PMID19931871
ABSTRACT
Selected POPs were analyzed in blubber samples of Pontoporia blainvillei from Southern Brazil to appraise temporal trend over a 10 year period (1994-2004). Overall, levels of POPs were relatively low, especially when compared to Northern Hemisphere concentrations. Apart from Mirex and PCBs, which showed stable concentrations, DDTs, HCB, CHLs, Dieldrin levels presented a slight decrease over the studied period. In addition, the increase in the PCBs/DDTs ratio supports the idea that inputs of DDTs are decaying faster than PCBs.