Merck
  • Developmental toxicity of diphenyl ether herbicides in nestling American kestrels.
Developmental toxicity of diphenyl ether herbicides in nestling American kestrels.
Journal of toxicology and environmental health (1991-11-01)
D J Hoffman, J W Spann, L J LeCaptain, C M Bunck, B A Rattner
ABSTRACT
Beginning the day after hatching, American kestrel (Falco sparverius) nestlings were orally dosed for 10 consecutive days with 5 microliters/g of corn oil (controls) or one of the diphenyl ether herbicides (nitrofen, bifenox, or oxyfluorfen) at concentrations of 10, 50, 250, or 500 mg/kg in corn oil. At 500 mg/kg, nitrofen resulted in complete nestling mortality, bifenox in high (66%) mortality, and oxyfluorfen in no mortality. Nitrofen at 250 mg/kg reduced nestling growth as reflected by decreased body weight, crown-rump length, and bone lengths including humerus, radius-ulna, femur, and tibiotarsus. Bifenox at 250 mg/kg had less effect on growth than nitrofen, but crown-rump, humerus, radius-ulna, and femur were significantly shorter than controls. Liver weight as a percent of body weight increased with 50 and 250 mg/kg nitrofen. Other manifestations of impending hepatotoxicity following nitrofen ingestion included increased hepatic GSH peroxidase activity in all nitrofen-treated groups, and increased plasma enzyme activities for ALT, AST, and LDH-L in the 250-mg/kg group. Bifenox ingestion resulted in increased hepatic GSH peroxidase activity in the 50- and 250-mg/kg groups. Nitrofen exposure also resulted in an increase in total plasma thyroxine (T4) concentration. These findings suggest that altricial nestlings are more sensitive to diphenyl ether herbicides than young or adult birds of precocial species.
Product Description
Supelco
Bifenox, PESTANAL®, analytical standard
Pricing and availability is not currently available.