- Fluorogenic assay for rapid detection of Escherichia coli in chilled and frozen foods: collaborative study.
Fluorogenic assay for rapid detection of Escherichia coli in chilled and frozen foods: collaborative study.
A collaborative study was conducted to compare a proposed LST-MUG method with the AOAC official method for Escherichia coli detection. E. coli produces an enzyme, beta-glucuronidase, which cleaves the substrate, 4-methyl-umbelliferyl-beta-D-glucuronide (MUG), to yield a fluorescent end product. Incorporation of the MUG substrate into lauryl tryptose broth (LST) enables a rapid quantitative method for screening E. coli, which is detected by fluorescence of the medium under longwave UV light. In this collaborative study, 5 food samples, 2 frozen (entree sauce/gravy and dairy topping) and 3 chilled (hamburger, pork sausage, and cheese), were tested for E. coli detection by 17 collaborating laboratories. Results indicate that the LST-MUG method is equal to or better than the current AOAC method for detecting E. coli. The LST-MUG method has been adopted official first action.