MilliporeSigma
  • Development of a novel high volume band compression injector for the analysis of complex samples like toxaphene pesticide.
Development of a novel high volume band compression injector for the analysis of complex samples like toxaphene pesticide.
Journal of chromatography. A (2008-10-22)
Jean-Pierre Gagné, Bruno Gouteux, Michel J Bertrand
ABSTRACT
A new type of injector has been developed for gas chromatographic analysis. The injector has high volume and band compression (HVBC) capabilities useful for the analysis of complex samples. The injector consists essentially of a packed liner operated at room temperature while a narrow heated zone is used to axially scan the liner selectively desorbing the compounds of interest. The scanning speed, distance and temperature of the zone are precisely controlled. The liner is connected to an interface which can vent the solvent or any undesirable compounds, and transfer the analytes to an analytical column for separation and quantification. The injector is designed to be compatible with injection volumes from 1 to more than 250microL. At a low sample volume of 1microL, the injector has competitive performances compared to those of the "on-column" and "split/splitless" injectors for the fatty acid methyl esters and toxaphene compounds tested. For higher volumes, the system produces a linear response according to the injected volume. In this explorative study, the maximum volume injected seems to be limited by the saturation of the chromatographic system instead of being defined by the design of the injector. The HVBC injector can also be used to conduct "in situ" pretreatment of the sample before its transfer to the analytical column. For instance, a toxaphene sample was successively fractionated, using the HVBC injector, in six sub-fractions characterized by simpler chromatograms than the chromatogram of the original mixture. Finally, the ability of the HVBC injector to "freeze" the separation in time allowing the analyst to complete the analysis at a later time is also discussed.
Product Description
Supelco
Toxaphene solution, certified reference material, 1000 μg/mL in isooctane