Merck
  • Practical, general, and systematic method for optical resolution of gem-dihalo- and monohalocyclopropanecarboxylic acids utilizing chiral 1,1'-binaphtholmonomethyl ethers: application to the synthesis of three chiral pesticides.
Practical, general, and systematic method for optical resolution of gem-dihalo- and monohalocyclopropanecarboxylic acids utilizing chiral 1,1'-binaphtholmonomethyl ethers: application to the synthesis of three chiral pesticides.
Organic & biomolecular chemistry (2008-01-26)
Hiroaki Yasukochi, Takayuki Atago, Akihiro Tanaka, Hidefumi Nakatsuji, Eri Yoshida, Akikazu Kakehi, Yoshinori Nishii, Yoo Tanabe
ABSTRACT
We performed an efficient practical and systematic optical resolution method for gem-dihalo- and monohalocyclopropanecarboxylic acids and utilizing chiral 1,1'-binaphthol monomethyl ether (R)- as the key auxiliary. Direct esterification of with (R)- gave two 1R- and 1S-diastereomeric esters with marked different R(f) values, both of which were easily separated using simple column chromatography. Monodehalogenation of separated chiral esters using t-BuMgCl and cat. Co(dppe)(2)Cl(2) gave two 1,2-trans- and 1,2-cis-diastereomers with markedly different R(f) values, both of which were similarly separated using simple column chromatography. The obtained diastereomers and were easily hydrolyzed to the desired enantiopure acids (>99%) and (>99%), respectively, with recovery of (R)-, both in good to excellent yields. Utilizing the present method, important chiral agrochemicals, carpropamid and fencyclate , were readily synthesized. Pyrethroid with three asymmetric centers was efficiently synthesized in a much better yield compared with the reported method.
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%