Merck
  • Development and application of oxygen production rate assessment to uptake of fluorinated organics by Lemna minor.
Development and application of oxygen production rate assessment to uptake of fluorinated organics by Lemna minor.
Environmental toxicology and chemistry (2006-11-09)
Dawn Marie Reinhold, E Michael Saunders
ABSTRACT
The effects of contaminant concentration on contaminant removal by Lemna minor were examined under elevated plant densities typical of full surface coverage. Uptake of 3-fluorophenol and 3-trifluoromethylphenol by L. minor was determined at concentrations from 10 to 1750 microM. Because standardized toxicity tests typically do not measure plant activity of L. minor at high plant densities, an oxygen production rate (OP) assessment was developed with consideration of experimental parameters that may inhibit OP by L. minor, including concentrations of carbonate and phosphate buffer in the media, time of assessment, and mass of L. minor. The OP by L. minor was inhibited by increasing media pH and mass of L. minor. The developed assessment used 0.5 g of L. minor per 60 ml of modified Standard Methods medium. A total of 30 h was needed to determine toxicity of fluorinated phenols, with an exposure period of 24 h, an incubation time of 6 h, and negligible analysis time. At concentrations from 10 to 1750 microM, 3-trifluoromethylphenol exhibited a sigmoidal toxicity response. However, 3-fluorophenol was neither toxic nor inhibitory at the experimental concentrations tested. Substantial decreases in uptake rate constants were observed for increasing concentrations of both contaminants, indicating that concentration may be a more important indicator of uptake by L. minor compared with decreasing plant activity because of toxicity.
Sigma-Aldrich
3-Fluorophenol, 98%
Pricing and availability is not currently available.