Merck
  • Serum 21-Deoxycortisol, 17-Hydroxyprogesterone, and 11-deoxycortisol in classic congenital adrenal hyperplasia: clinical and hormonal correlations and identification of patients with 11beta-hydroxylase deficiency among a large group with alleged 21-hydroxylase deficiency.
Serum 21-Deoxycortisol, 17-Hydroxyprogesterone, and 11-deoxycortisol in classic congenital adrenal hyperplasia: clinical and hormonal correlations and identification of patients with 11beta-hydroxylase deficiency among a large group with alleged 21-hydroxylase deficiency.
The Journal of clinical endocrinology and metabolism (2006-03-23)
Vania Tonetto-Fernandes, Sofia H V Lemos-Marini, Hilton Kuperman, Luciane M Ribeiro-Neto, Ieda T N Verreschi, Claudio E Kater
ABSTRACT
21-Hydroxylase deficiency (21OHD) is the most common cause of congenital adrenal hyperplasia, followed in frequency by 11beta-hydroxylase deficiency (11betaOHD). Although the relative frequency of 11betaOHD is reported as between 3 and 5% of the cases, these numbers may have been somewhat underestimated. In 133 patients (89 females/44 males; 10 d-20.9 yr) with alleged classic 21OHD and five (three females/two males; 7.3-21 yr) with documented 11betaOHD, we measured serum 21-deoxycortisol (21DF), 17-hydroxyprogesterone (17OHP), and 11-deoxycortisol (S), 48 h after glucocorticoid withdrawal. We also studied 20 sex- and age-matched control subjects. Serum steroid levels were determined by RIA after HPLC purification. The objectives of this study were to: 1) quantify 21DF in patients with congenital adrenal hyperplasia, 2) correlate hormonal with clinical data, and 3) identify possible misdiagnosed patients with 11betaOHD among those with 21OHD. In 21OHD, 17OHP (217-100,472 ng/dl) and 21DF (<39-14,105 ng/dl) were mostly elevated and positively correlated (r = 0.7202; P < 0.001). Except for higher 17OHP in pubertal patients, 17OHP and 21DF values were similar according to sex, disease severity, or prevailing glucocorticoid dose. One additional patient with 11betaOHD was detected (1%) and also one with apparent combined 11beta- and 21OHD. S levels were elevated in 11betaOHD and normal but significantly higher in 21OHD than in controls. To recognize patients with 21- and/or 11betaOHD, we recommend evaluation of 17OHP or 21DF and S. Also, 21DF may be useful to follow up pubertal patients with 21OHD. Because 1% of patients with alleged 21OHD may have 11betaOHD, its frequency seems underestimated, as per our experience in a Brazilian population.
Product Description
Supelco
21-Deoxycortisol solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
21-Deoxycortisol
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%
Supelco
17α-Hydroxyprogesterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®