- Management of psychosis in Australian general practice.
Management of psychosis in Australian general practice.
Australian family physician (2006-03-10)
Janice Charles, Graeme Miller, Anthea Ng
PMID16525516
ABSTRACT
The BEACH program, a continuous national study of general practice activity in Australia, gives us an overview of consultations involving the management of psychoses. In this analysis we have included schizophrenia, affective disorders/bipolar, organic psychoses, and senile psychoses, with undefined psychosis and chronic brain syndrome grouped as 'other'. This synopsis provides a backdrop against which the theme articles in this issue of Australian Family Physician can be further considered.