Merck
  • Sperm mobility: deduction of a model explaining phenotypic variation in roosters (Gallus domesticus).
Sperm mobility: deduction of a model explaining phenotypic variation in roosters (Gallus domesticus).
Biology of reproduction (2005-11-18)
D P Froman, J C Wardell, A J Feltmann
ABSTRACT
In previous work, variation in sperm mobility phenotype was attributed to the proportion of ejaculated fowl sperm containing dysfunctional mitochondria. In the present work, latent mitochondrial dysfunction was inferred from patterns of sperm egress from the oviduct's sperm-storage tubules. In addition, experiments were performed to help explain how mitochondrial function could be compromised in viable sperm cells. Confocal microscopy demonstrated that sperm Ca2+ content differed between low and high sperm-mobility phenotypes when sperm were stained with rhod-2 AM, a Ca2+ -specific dye. Fluorescence was associated with the nuclear envelope, a variant of the endoplasmic reticulum, and greater fluorescence was observed in sperm from low sperm-mobility males. Fluorescence was reduced by 50% when motile sperm were rendered immotile by incubation with a Ca2+ chelator. Thus, a relationship was established between a dynamic intracellular Ca2+ pool and sperm motility. Sperm N-methy-D-aspartic acid (NMDA) receptors were inferred by the action of D-homocysteinesulfinic acid, a potent NMDA receptor agonist. Seminal plasma from low sperm mobility males was characterized by an elevated glutamate concentration. Thapsigargin, which inhibits the smooth endoplasmic reticulum Ca2+ pump and thereby promotes Ca2+ efflux, rendered sperm immotile. This effect was blocked by cyclosporin A, which prevents the formation of the mitochondrial permeability transition pore (PTP) in response to elevated mitochondrial Ca2+ content. In summary, we propose that 1) glutamate enables Ca2+ uptake into sperm before ejaculation, 2) excessive Ca2+ uptake triggers formation of the PTP in a subpopulation of sperm, and 3) sperm mobility is decreased in proportion.