Merck
  • Insight into pollutant bioavailability and toxicity using Raman confocal microscopy.
Insight into pollutant bioavailability and toxicity using Raman confocal microscopy.
Journal of microbiological methods (2005-01-15)
Andrew C Singer, Wei E Huang, Jana Helm, Ian P Thompson
ABSTRACT
Raman confocal microscopy was used to discriminate between cultures of Burkholderia xenovorans LB400 exposed to four different common environmental pollutants: phenanthrene, dodecane, 3-chlorobiphenyl and pentachlorophenol. Evidence is presented for the application of Raman spectroscopy as a bioassay for pollutant bioavailability and toxicity.