Merck
  • Synthesis and characterization of poly(dimer acid-brassylic acid) copolymer and poly(dimer acid-pentadecandioic acid) copolymer.
Synthesis and characterization of poly(dimer acid-brassylic acid) copolymer and poly(dimer acid-pentadecandioic acid) copolymer.
Biopolymers (2004-05-20)
Wen-xun Guo, Kai-xun Huang
ABSTRACT
Poly(dimer acid-brassylic acid) [P(DA-BA)] copolymers and poly(dimer acid-pentadecandioic acid) [P(DA-PA)] copolymers were prepared by melt polycondensation of the corresponding mixed anhydride prepolymers. The copolymers were characterized by Fourier transform infrared (FTIR), gel permeation chromatography (GPC), differential scanning calorimetry (DSC), wide angle x-ray powder-diffraction, and thermal gravimetric analysis (TGA). In vitro studies show that all the copolymers are degradable in phosphate buffer at 37 degrees C, and leaving an oily dimer acid residue after hydrolysis for the copolymer with high content of dimer acid. The release profiles of hydrophilic model drug, ciprofloxcin hydrochloride, from the copolymers, follow first-order release kinetics. All the preliminary results suggested that the copolymer might be potentially used as drug delivery devices.