Merck
  • Assessing gene expression in lung subcompartments utilizing in situ RNA preservation.
Assessing gene expression in lung subcompartments utilizing in situ RNA preservation.
Toxicological sciences : an official journal of the Society of Toxicology (2003-11-06)
Gregory L Baker, Michael A Shultz, Michelle V Fanucchi, Dexter M Morin, Alan R Buckpitt, Charles G Plopper
ABSTRACT
The mechanisms of toxicant-mediated lung injury and repair are influenced by the considerable spatial heterogeneity that exists within the conducting airways of the lungs. As a result of this heterogeneity, significant differences and similarities in gene expression are observed throughout lung subcompartments. RNA-based technologies such as real-time reverse transcription polymerase chain reaction (real-time RT-PCR) and cDNA microarray analysis of gene expression provide valuable clues to understanding the mechanisms of toxicant-induced injury. Isolating RNA from lung subcompartments has previously involved considerable time and labor-intensive processes that limit the number of animals that could be processed in a day. The aim of this study was to determine if intact, high-quality RNA could be preserved in situ over a period of time to delay the need to immediately perform site-specific lung subcompartment microdissections and RNA isolations. Two hours after 1-nitronaphthalene treatment, rat lungs were inflated with and stored in RNA preservation solution and stored at 4 degrees C for 7 days. RNA was isolated from the lung subcompartments isolated by microdissection. After 7 days of storage, the RNA was intact, of high quality, and could be used for real-time RT-PCR to examine heterogeneous gene expression in the lung subcompartments. In summary, this simplified technique of in situ RNA preservation and site-specific lung subcompartment microdissection allows the isolation of intact, high-quality RNA that may be used with molecular RNA-based technologies that will significantly accelerate our understanding of pulmonary injury and repair mechanisms.
Product Description
1-Nitronaphthalene, BCR®, certified reference material