Merck
  • Influence of agents affecting voltage-dependent calcium channels and dantrolene on the anticonvulsant action of the AMPA/kainate receptor antagonist LY 300164 in mice.
Influence of agents affecting voltage-dependent calcium channels and dantrolene on the anticonvulsant action of the AMPA/kainate receptor antagonist LY 300164 in mice.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (2002-07-20)
Mariusz Swiader, Kinga K Borowicz, Jacek Porebiak, Zdzisław Kleinrok, Stanisław J Czuczwar
ABSTRACT
It was previously documented that calcium (Ca(2+)) channel inhibitors intensified the protective effects of conventional antiepileptics against electroconvulsions in mice. The aim of this study was to evaluate the effects of Ca(2+) channel inhibitors (nifedipine, nicardipine and flunarizine) on the anticonvulsant action of the new AMPA/kainate receptor antagonist, 7-acetyl-3-(4-aminophenyl)-8,9-dihydro-8-methyl-7H-1,3-dioxazolo[4,5-h][2,3]-benzodiazepine (LY 300164), against maximal electroshock (MES)-induced seizures in mice. Dantrolene (an inhibitor of Ca(2+)release from intracellular stores) was also included. Nifedipine (30 mg/kg) and flunarizine (15 mg/kg) raised the threshold for electroconvulsions, being ineffective at lower doses. Nicardipine (up to 30 mg/kg) and dantrolene (up to 20 mg/kg) did not affect this parameter. Flunarizine (10 mg/kg), nicardipine (20 mg/kg) and dantrolene (20 mg/kg) potentiated the efficacy of LY 300164 against MES. However, nicardipine (at 20 mg/kg) raised the free plasma concentration of LY 300164. Nifedipine (30 mg/kg), given even in a dose raising the electroconvulsive threshold, did not significantly alter the protective effect of LY 300164 against MES. Furthermore, the Ca(2+) channel agonist-BAY k-8644 (at 5 mg/kg) did not influence the protection offered by LY 300164 against MES. Finally, this Ca(2+) channel activator did not affect the enhanced efficacy of LY 300164 by Ca(2+) channel modulators. The only exception was the combination of LY 300164 with flunarizine. Combined treatment with LY 300164 and dantrolene (20 mg/kg), compared to LY 300164 alone, resulted in an impairment of motor performance in mice. Ca(2+) channel inhibitors were without effect upon this parameter evaluated in the chimney test. As shown in the passive avoidance task, LY 300164 alone (at its ED(50)) or combined with agents affecting neuronal Ca(2+) concentration did not disturb long-term memory. The present results suggest that agents preventing influx of Ca(2+) ions into neurons may enhance the protective action of LY 300164.