Merck
  • The systems V(IV)O(2+)-glutathione and related ligands: a potentiometric and spectroscopic study.
The systems V(IV)O(2+)-glutathione and related ligands: a potentiometric and spectroscopic study.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2002-04-06)
João Costa Pessoa, Isabel Tomaz, Tamás Kiss, Erzsébet Kiss, Péter Buglyó
ABSTRACT
The equilibria in the system V(IV)O(2+)-glutathione in aqueous solution were studied in the pH range 2-11 by a combination of pH-potentiometry and spectroscopy (EPR, visible absorption and circular dichroism). The results of the various methods are consistent and the equilibrium model includes the species MLH(3), MLH(2), MLH, ML(2)H(2), MLH(-1), and MLH(-2) and several hydrolysis products (where H(4)L denotes totally protonated glutathione); individual formation constants and spectra are given. ML(2)H(2) is the predominant species at physiological pH. Plausible structures for each stoichiometry are discussed. The related V(IV)O(2+) systems of S-methylglutathione and gamma- L-glutamyl- L-cysteinyl ethyl ester were studied by means of the same spectroscopic techniques in order to support the established binding modes for the glutathione complexes. The importance of glutathione and oxidized glutathione in binding V(IV)O(2+) in cells is assessed.
Sigma-Aldrich
S-Methylglutathione