Merck
Vibrational frequencies and structural determinations of 1,4-thioxane.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-01-05)
M W Ellzy, J O Jensen, H F Hameka, J G Kay, D Zeroka
ABSTRACT
We present a detailed analysis of the structure and infrared spectra of 1,4-thioxane. The vibrational frequencies of the 1,4-thioxane molecule were analyzed using standard quantum chemical techniques. Frequencies were calculated at the MP2 and DFT levels of theory using the standard 6-31G* basis set. The structural transformation of the chemical agent bis (2-chloroethyl) sulfide (HD, mustard gas) and the related symmetry to a previously study compound(4) makes the symmetry of the 1,4-thioxane molecule an interesting candidate for study. The molecule exists normally in a Cs configuration similar to the chair form of cyclohexane. High-energy forms of 1,4-thioxane with C1 and C2 symmetry also exist.
Sigma-Aldrich
1,4-Thioxane, 98%
Pricing and availability is not currently available.