Merck
  • Deposition of 7-aminoflunitrazepam and flunitrazepam in hair after a single dose of Rohypnol.
Deposition of 7-aminoflunitrazepam and flunitrazepam in hair after a single dose of Rohypnol.
Journal of forensic sciences (2001-09-25)
A Negrusz, C M Moore, K B Hinkel, T L Stockham, M Verma, M J Strong, P G Janicak
ABSTRACT
In recent years, there has been a notable increase in the number of reports on drug-facilitated sexual assault. Benzodiazepines are the most common so-called "date-rape" drugs, with flunitrazepam (Rohypnol) being one of the most frequently mentioned. The aim of this study was to determine whether flunitrazepam and its major metabolite 7-aminoflunitrazepam could be detected in hair collected from ten healthy volunteers after receiving a single 2 mg dose of Rohypnol using solid phase extraction and NCI-GC-MS. Such data would be of great importance to law enforcement agencies trying to determine the best time interval for hair collection from a victim of drug-facilitated sexual assault in order to reveal drug use. Ten healthy volunteers (eight women and two men, 21 to 49 years old) participated in the study. The following hair samples were collected from each volunteer: one before flunitrazepam administration, and 1, 3, 5, 14, 21, and 28 days after. In five volunteers, 7-aminoflunitrazepam was detected 24 h after flunitrazepam administration and remained in hair throughout the entire 28-day study period (0.6-8.0 pg/mg). In two cases, 7-aminoflunitrazepam appeared in hair 21 days after drug intake (0.5-2.7 pg/mg), and in two subjects 14 days later (0.5-5.4 pg/mg). In one volunteer, 7-aminoflunitrazepam was detected on day 14 and 21 but concentrations were below the quantitation limit. Flunitrazepam was detected in some samples but all concentrations were below the quantitation limit (0.5-2.3 pg/mg).
Product Description
Supelco
7-Aminoflunitrazepam solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®