Merck
  • A heterocycle-forming double michael reaction. [5 + 1] annulation route to highly substituted and functionalized piperidines.
A heterocycle-forming double michael reaction. [5 + 1] annulation route to highly substituted and functionalized piperidines.
Organic letters (2001-09-01)
F Hughes, R B Grossman
ABSTRACT
[reaction: see text]. Nitrogen-containing tethered diacids, easily prepared by reductive alkylation of diethyl aminomalonate or ethyl cyanoglycinate, undergo double Michael reactions with 3-butyn-2-one to give highly functionalized and substituted piperidines (pipecolic acid derivatives) with surprisingly high stereoselectivity. The heterocyclic double Michael adducts can be induced to undergo further cyclizations to give a variety of azabicyclic and diazabicyclic compounds.
Sigma-Aldrich
Diethyl aminomalonate hydrochloride, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
3-Butyn-2-one, 95%
Pricing and availability is not currently available.