Merck
  • Non-hydrogen-bonded secondary structure in beta-peptides: evidence from circular dichroism of (S)-pyrrolidine-3-carboxylic acid oligomers and (S)-nipecotic acid oligomers.
Non-hydrogen-bonded secondary structure in beta-peptides: evidence from circular dichroism of (S)-pyrrolidine-3-carboxylic acid oligomers and (S)-nipecotic acid oligomers.
Organic letters (2000-06-03)
B R Huck, J M Langenhan, S H Gellman
ABSTRACT
[formula: see text] Homooligomers of beta-amino acids (S)-3-pyrrolidine-3-carboxylic acid (PCA) and (S)-nipecotic acid (Nip) were studied by circular dichroism (CD) in methanol. In each series, a profound change in the far-UV CD spectrum was observed from monomer to tetramer, but little change was observed from tetramer to hexamer. A comparable pattern is observed in the CD spectra of short proline oligomers. We conclude that both PCA and Nip oligomers with > or = four residues adopt a characteristic secondary structure.
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)
Pricing and availability is not currently available.