Merck
  • Triplet-state energies and substituent effects of excited aroyl compounds in the gas phase.
Triplet-state energies and substituent effects of excited aroyl compounds in the gas phase.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2000-03-23)
Z P Lin, W A Aue
ABSTRACT
Triplet-state energy values obtained from the gas phase are still scarce. In this study, the triplet-state energies of 58 aroyl compounds, introduced as gas chromatographic peaks at atmospheric pressure and typically 473 K, have been determined from the 0-0 bands of their n --> pi* type phosphorescence spectra in excited nitrogen. Correlations of those gas-phase triplet-state energies with Hammett constants could be observed for substituted acetophenones, benzaldehydes and benzophenones.
Sigma-Aldrich
m-Tolualdehyde, 97%