Merck
  • Penitricin, a new class of antibiotic produced by Penicillium aculeatum. II. Isolation and characterization.
Penitricin, a new class of antibiotic produced by Penicillium aculeatum. II. Isolation and characterization.
The Journal of antibiotics (1984-07-01)
T Okuda, K Yokose, T Furumai, H B Maruyama
ABSTRACT
A novel antibiotic, penitricin, Ro 09-0804, has been produced in the culture filtrate of Penicillium aculeatum NR 5165. This antibiotic was purified by repeated extraction of culture filtrate with 1-butanol, and passage of the crude extracts through Sephadex G-10, followed by HPLC (Shodex Ionpak S-801). Physico-chemical characterization was made on penitricin, while two open-ringed penitricins B and C co-produced by the producer strain were also identified.
Sigma-Aldrich
cis-2-Butene-1,4-diol, 97%