Merck
  • Relaxivity control of magnetic nanoclusters for efficient magnetic relaxation switching assay.
Relaxivity control of magnetic nanoclusters for efficient magnetic relaxation switching assay.
Chemical communications (Cambridge, England) (2012-10-27)
Jinmyung Cha, Yong-Su Kwon, Tae-Jong Yoon, Jin-Kyu Lee
ABSTRACT
50 nm Magnetic nanocluster (MNC) was deliberately selected as an efficient probe for the magnetic relaxation switching (MRSw) assay. After surface modification of MNCs with poly(acrylic acid) to give PAA-MNCs, and employing streptavidin-biotin interactions for the MRSw assay model, a few tenths pM concentration of strepavidin could be determined.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis