Merck
  • Involvement of histamine receptors in the atypical antipsychotic profile of clozapine: a reassessment in vitro and in vivo.
Involvement of histamine receptors in the atypical antipsychotic profile of clozapine: a reassessment in vitro and in vivo.
Psychopharmacology (2011-09-14)
Marie Humbert-Claude, Elisabeth Davenas, Florence Gbahou, Ludwig Vincent, Jean-Michel Arrang
ABSTRACT
The basis of the unique clinical profile of the antipsychotic clozapine is not yet elucidated. Brain histamine receptors may play a role in schizophrenia and its treatment, but their involvement in the profile of clozapine remained unknown. We explored the properties of clozapine and its two metabolites, N-desmethylclozapine (NDMC) and clozapine N-oxide, at the four human histaminergic receptors. We compared their active concentrations with their blood concentrations in patients treated by clozapine. We investigated the changes in receptor densities induced in rat brain by repeated administration of a therapeutic dose of clozapine. Clozapine and NDMC behaved as very potent, and partial, H(1)-receptor inverse agonists, weak, and full, H(2)-receptor inverse agonists, moderate, and protean, H(3)-receptor agonists, and moderate, and partial, H(4)-receptor agonists. Taking into account their micromolar mean blood concentrations found in 75 treated patients, and assuming that they are enriched in human brain as they are in rat brain, a full occupation of H(1)-, H(3)-, and H(4)-receptors, and a partial occupation of H(2) receptors, is expected. In agreement, repeated administration of clozapine at a therapeutic dose (20 mg/kg/day for 20 days) induced an up-regulation of H(1)- and H(2)-receptors in rat brain. Clozapine and its active metabolite NDMC interact with the four human histamine receptors at clinically relevant concentrations. This interaction may substantiate, at least in part, the atypical antipsychotic profile of clozapine, as well as its central and peripheral side effects such as sedation and weight gain.
Product Description
Supelco
N-Desmethylclozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®