Merck
  • Biotransformation of ginsenoside Rf to Rh1 by recombinant beta-glucosidase.
Biotransformation of ginsenoside Rf to Rh1 by recombinant beta-glucosidase.
Molecules (Basel, Switzerland) (2009-06-11)
Chang-Chun Ruan, Hao Zhang, Lian-Xue Zhang, Zhi Liu, Guang-Zhi Sun, Jun Lei, Yu-Xia Qin, Yi-Nan Zheng, Xiang Li, Hong-Yu Pan
ABSTRACT
An Aspergillus niger strain was isolated from the soil around ginseng fruit. In vitro enzyme assays showed that this strain had the ability to transform total ginsenosides (TGS) into several new products. In a further biochemical study, a beta-glucosidase gene isolated from this strain, bgl1, was expressed in Saccharomyces cerevisiae. His-tagged BGL1 protein (approximately 170 kD) showed the ability to transform ginsenoside Rf into Rh(1).
Product Description
Supelco
Ginsenoside Rf, analytical standard
Ginsenoside Rf, primary reference standard
Supelco
Ginsenoside Rh1, analytical standard