Merck
  • A fast and inexpensive method for N-terminal fluorescein-labeling of peptides.
A fast and inexpensive method for N-terminal fluorescein-labeling of peptides.
Bioorganic & medicinal chemistry letters (1999-01-01)
P J Weber, J E Bader, G Folkers, A G Beck-Sickinger
ABSTRACT
A new method has been developed to synthesize fluorescein labeled peptides, compounds of increasing importance in bioorganic chemistry, cell biology, pharmacology, drug targeting and medicinal chemistry. We show, that 4(5)-carboxyfluorescein is much more efficient than the hitherto predominantly utilized reagents 4(5)-carboxyfluorescein-N-succinimidylester and 4(5)-fluoresceinisothiocyanate.
Sigma-Aldrich
DIC, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
DIC, purum, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF
Pricing and availability is not currently available.