Merck
  • Deodorants on the European market: quantitative chemical analysis of 21 fragrances.
Deodorants on the European market: quantitative chemical analysis of 21 fragrances.
Contact dermatitis (1998-03-21)
S C Rastogi, J D Johansen, P Frosch, T Menné, M Bruze, J P Lepoittevin, B Dreier, K E Andersen, I R White
ABSTRACT
Deodorants are one of the most frequently used types of cosmetics and side-effects from them are common. Recent studies relate perfume allergy to this type of product. 73 deodorants were analyzed by gas chromatography--mass spectrometry for the determination of the contents of 7 wellknown fragrance allergens from the fragrance mix and 14 other commonly used fragrance materials. The deodorants were purchased at retail outlets in 5 European countries. It was found that in general, fragrance mix ingredients were more frequently present in vapo- and aerosol sprays than in roll-on products. The levels of the fragrance mix substances ranged from 0.0001-0.2355%. The products investigated contained cinnamic aldehyde and isoeugenol less frequently (17% and 29% respectively), and eugenol and geraniol most frequently (57% and 76% respectively). The 14 other fragrance materials were found in 40-97% of the deodorants, with hedione and benzyl acetate the most frequently found substances. The concentration of these 14 substances ranged from 0.0001-2.7%. It is concluded that the levels of cinnamic aldehyde and isoeugenol found in the deodorants could prove to be relevant for elicitation of contact dermatitis. No conclusions could be drawn about the other fragrance mix constituents, as threshold levels in sensitized individuals have not been investigated. Furthermore, all of the fragrance materials investigated were frequently found in deodorants and, apart from the fragrance mix ingredients, the extent of problems with sensitization to these fragrance materials is largely unknown.
Sigma-Aldrich
α-Amylcinnamaldehyde, mixture of cis and trans, ≥97%, stabilized, FG
Pricing and availability is not currently available.
Sigma-Aldrich
α-Amylcinnamaldehyde, 97%, mixture of cis and trans
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
7-Hydroxycitronellal, Odorant used in allergy studies
Pricing and availability is not currently available.