Merck
  • Analysis of trifluoroacetic acid in lyophilized natural products by capillary electrophoresis.
Analysis of trifluoroacetic acid in lyophilized natural products by capillary electrophoresis.
Journal of chromatography. B, Biomedical sciences and applications (1997-10-29)
M A Strege, W G Mascher
ABSTRACT
A rapid and simple method for the capillary electrophoretic determination of residual trifluoroacetic acid in lyophilized natural products is described. The technique utilizes 2,6-naphthalenedicarboxylic acid as a separation buffer additive providing indirect ultraviolet absorption detection. Using this method, acceptable precision, accuracy, selectivity, range and linearity were achieved.
Sigma-Aldrich
2,6-Naphthalenedicarboxylic acid, 95%
Sigma-Aldrich
2,6-Naphthalenedicarboxylic acid, 99%