Merck
  • The solid-phase synthesis of side-chain-phosphorylated peptide-4-nitroanilides.
The solid-phase synthesis of side-chain-phosphorylated peptide-4-nitroanilides.
The journal of peptide research : official journal of the American Peptide Society (1997-08-01)
A Bernhardt, M Drewello, M Schutkowski
ABSTRACT
Peptide-4-nitroanilides can be quickly synthesised using an Fmoc-based approach on 2-chlorotritylchloride resin. Preformed building blocks Fmoc-Xaa-NH-Np (Xaa = Cit, Cys, Gln, His, Lys, Orn, Ser, Thr, Tyr, Trp) can be attached via side chain to the 2-chlorotritylchloride linker of the resin. N-terminal elongation yields the respective peptide-4-nitroanilides after detachment from the solid support. We synthesised a set of tetrapeptide-4-nitroanilides with the general structure Suc-Ala-Phe-Pro-Xaa-NH-Np (Xaa = Asp, Cit, Cys, Glu, Gln, His, Lys, Orn, Ser, Thr, Tyr, Trp). Even peptidyl-arginine-4-nitroanilides are available by a slightly modified procedure. First, the appropriate ornithine-containing peptide was synthesised. After detachment of the peptide from the resin the side-chain primary amino group was transformed to the guanidino function of arginine using 1-guanyl-3,5-dimethylpyrazole. A further application of this method is the convenient synthesis of phosphorylated peptide-4-nitroanilides. Five phosphopeptides with the general structure Ac-Ala-Xaa(PO3H2)-Pro-Yaa-NH-Np (Xaa = Ser, Thr, Tyr; Yaa = Tyr, Lys) and their nonphosphorylated analogues were prepared. Global phosphorylation was carried out on the resin-bound peptides using dibenzyl-N, N-diisopropyl-phosphoramidite/tetrazole followed by oxidation with tert-butyl hydroperoxide.
Sigma-Aldrich
3,5-Dimethyl-1-pyrazolylformaminidium nitrate, 97%
Pricing and availability is not currently available.