Merck
  • Production of useful chemicals from sunflower oil by microbial biotransformation with rhizopus stolonifer NRRL 1478.
Production of useful chemicals from sunflower oil by microbial biotransformation with rhizopus stolonifer NRRL 1478.
Australasian biotechnology (1996-01-01)
S H El-Sharkawy
ABSTRACT
A number of microorganisms has been screened for growth on sunflower oil as a sole carbon source for production of useful chemicals. Rhizopus stolonifer NRRL 1478 was found to transform the lipid contents of sunflower oil into dodecyl b-D-glucopyranoside and dodecanedioic acid in 15 and 25% yields respectively. The produced compounds were isolated and purified by column chromatography and their chemical identities were established using MS, IR, 1H and 13C NMR spectroscopy.
Sigma-Aldrich
n-Dodecyl β-D-glucopyranoside, ≥98% (GC)