Merck
  • Identification of new polymorphisms of the angiotensin I-converting enzyme (ACE) gene, and study of their relationship to plasma ACE levels by two-QTL segregation-linkage analysis.
Identification of new polymorphisms of the angiotensin I-converting enzyme (ACE) gene, and study of their relationship to plasma ACE levels by two-QTL segregation-linkage analysis.
American journal of human genetics (1996-06-01)
E Villard, L Tiret, S Visvikis, R Rakotovao, F Cambien, F Soubrier
ABSTRACT
Plasma angiotensin I-converting enzyme (ACE) levels are highly genetically determined. A previous segregation-linkage analysis suggested the existence of a functional mutation located within or close to the ACE locus, in almost complete linkage desequilibrium (LD) with the ACE insertion/deletion (I/D) polymorphism and accounting for half the ACE variance. In order to identify the functional variant at the molecular level, we compared ACE gene sequences between four subjects selected for having contrasted ACE levels and I/D genotypes. We identified 10 new polymorphisms, among which 8 were genotyped in 95 healthy nuclear families, in addition to the I/D polymorphism. These polymorphisms could be divided into two groups: five polymorphisms in the 5' region and three in the coding sequence and the 3' UTR. Within each group, polymorphisms were in nearly complete association, whereas polymorphisms from the two groups were in strong negative LD. After adjustment for the I/D polymorphism, all polymorphisms of the 5' group remained significantly associated with ACE levels, which suggests the existence of two quantitative trait loci (QTL) acting additively on ACE levels. Segregation-linkage analyses including one or two ACE-linked QTLs in LD with two ACE markers were performed to test this hypothesis. The two QTLs and the two markers were assumed to be in complete LD. Results supported the existence of two ACE-linked QTLs, which would explain 38% and 49% of the ACE variance in parents and offspring, respectively. One of these QTLs might be the I/D polymorphism itself or the newly characterized 4656(CT)2/3 polymorphism. The second QTL would have a frequency of approximately .20, which is incompatible with any of the yet-identified polymorphisms. More extensive sequencing and extended analyses in larger samples and in other populations will be necessary to characterize definitely the functional variants.
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly
Pricing and availability is not currently available.