Merck
  • Biological fate of sulphur mustard, 1,1'-thiobis(2-chloroethane): identification of beta-lyase metabolites and hydrolysis products in human urine.
Biological fate of sulphur mustard, 1,1'-thiobis(2-chloroethane): identification of beta-lyase metabolites and hydrolysis products in human urine.
Xenobiotica; the fate of foreign compounds in biological systems (1995-02-01)
R M Black, R W Read
ABSTRACT
1. Samples of urine from two human subjects accidentally exposed to sulphur mustard were analysed for metabolites derived from hydrolysis (thiodiglycol, thiodiglycol sulphoxide), conjugation with glutathione (1,1'-sulphonylbis [2-S-(N-acetylcysteinyl)ethane]) and from further metabolism of glutathione conjugates by the beta-lyase pathway (1,1-sulphonylbis[2-(methylsulphinyl)ethane], 1-methylsulphinyl-2-[2-(methylthio)ethylsulphonyl]ethane). 2. Thiodiglycol sulphoxide was excreted in much higher concentrations than thiodiglycol, as was observed previously in rat exposed to sulphur mustard. However, the use of thiodiglycol sulphoxide as a biological marker for sulphur mustard poisoning is limited by its presence at low concentrations in normal human urine. 3. beta-lyase metabolites were detected at concentrations comparable with those of thiodiglycol sulphoxide. No background levels of beta-lyase metabolites have been detected in normal human urine, and they are proposed as unequivocal diagnostic and forensic indicators of sulphur mustard poisoning in man.
Sigma-Aldrich
2,2′-Sulfonyldiethanol solution, 60-65 wt. % in H2O
Pricing and availability is not currently available.