Merck
  • [Synthesis of New Uracil Derivatives / On the reactability of 4-chloromethyluracil (author's transl)].
[Synthesis of New Uracil Derivatives / On the reactability of 4-chloromethyluracil (author's transl)].
Arzneimittel-Forschung (1980-01-01)
J Klosa
ABSTRACT
The synthesis of new uracil derivatives is described. In 4-chloromethyluracil, chlorine can be easily exchanged under mild conditions for amine, aniline, hydrazine, and phenol.
Sigma-Aldrich
6-(Chloromethyl)uracil, 98%
Pricing and availability is not currently available.