Merck
  • Quantification of amitriptyline, nortriptyline, and 10-hydroxy metabolite isomers in plasma by capillary gas chromatography with nitrogen-sensitive detection.
Quantification of amitriptyline, nortriptyline, and 10-hydroxy metabolite isomers in plasma by capillary gas chromatography with nitrogen-sensitive detection.
Journal of chromatography (1983-12-09)
D R Jones, B J Lukey, H E Hurst
ABSTRACT
A selective, sensitive method for the determination of amitriptyline and its metabolites is described. This method involves liquid-liquid extraction and capillary gas chromatography with nitrogen-sensitive detection. The detection limits of amitriptyline, nortriptyline, 10-hydroxy(E)amitriptyline, 10-hydroxy(E)nortriptyline, and 10-hydroxy(Z)nortriptyline were slightly less than 0.5 ng/ml in 1.0-ml plasma samples. The coefficients of variation for within-run and between-run analyses of samples containing 100 ng/ml were less than 12% and 9%, respectively. The method offers rapid analysis of individual isomers, increased sensitivity over high-performance liquid chromatographic methodology and the conveniences of the gas chromatographic technique.