Merck
Human maximization testing of D&C Yellow no. 10 and Yellow no. 11.
Contact dermatitis (1984-10-01)
S Kita, T Kobayashi, H Kutsuna, A M Kligman
ABSTRACT
By the maximization test, using 0.5% D&C Yellow no. 11 in pet., 15 of 20 human volunteers became contact sensitized. All reacted to a challenge concentration of 1000 ppm and one down to 1 ppm. The high allergenic potential of this dyestuff was confirmed. There were no cross reactions to purified samples of D&C Yellow no. 10. In contrast, 5% solutions of pure D&C Yellow no. 10 failed to sensitize human subjects. Cross reactions to this substance in persons sensitized to D&C Yellow no. 11 are due to impurities.
Sigma-Aldrich
Quinoline Yellow, Dye content 95 %
Sigma-Aldrich
Quinoline Yellow, for microscopy (Hist.), mixture of mono- and disulfonic acid sodium salt
Sigma-Aldrich
Quinoline Yellow, Mixture of the mono- and disulfonic acids of Quinoline Yellow