Merck
  • Effect of the threonine analog beta-hydroxynorvaline on the glycosylation and secretion of alpha 1-acid glycoprotein by rat hepatocytes.
Effect of the threonine analog beta-hydroxynorvaline on the glycosylation and secretion of alpha 1-acid glycoprotein by rat hepatocytes.
The Journal of biological chemistry (1985-09-05)
P A Docherty, N N Aronson
ABSTRACT
The threonine analog beta-hydroxynorvaline (Hnv) is an inhibitor of asparagine-linked glycosylation. In the presence of the analog hepatocytes synthesized immunoreactive alpha 1-acid glycoprotein with 0-6 oligosaccharide chains. Pulse-chase experiments were conducted to compare the rates of secretion of alpha 1-acid glycoprotein from untreated, tunicamycin-treated, and Hnv-treated cells. Partially glycosylated (1-5 oligosaccharide chains) and unglycosylated (tunicamycin-inhibited) molecules exited the cells more slowly than native alpha 1-acid glycoprotein. In addition, secretion of fully glycosylated (6 oligosaccharide chains) alpha 1-acid glycoprotein was retarded in Hnv-treated cells when compared to controls. The slowest rate of secretion was exhibited by the unglycosylated form from Hnv-treated cells. These results suggest that Hnv-induced changes either in the extent of glycosylation or in the peptide sequence of alpha 1-acid glycoprotein can interfere with its transport through the cell. The major intracellular forms of alpha 1-acid glycoprotein from control and Hnv-treated cells were endoglycosidase H-sensitive and contained Man9-8 GlcNAc2 oligosaccharide structures. The oligosaccharide chains on the secreted molecules from control and Hnv-treated cells were entirely of the endoglycosidase H-resistant, complex type.
Sigma-Aldrich
DL-3-Hydroxynorvaline, ≥98% (TLC)