Merck
  • The effect of thai glutinous rice starch on the synthesis of lead zirconate (PbZrO3) nanofibers via the electrospinning method.
The effect of thai glutinous rice starch on the synthesis of lead zirconate (PbZrO3) nanofibers via the electrospinning method.
Journal of nanoscience and nanotechnology (2013-07-19)
Chanisa Nawanil, Wanwilai Vittayakorn, Rangson Muanghua, Surasak Niemcharoen, Jutarat Prachayawarakorn, Naratip Vittayakorn
ABSTRACT
This study synthesized blended lead zirconate (PbZrO3;PZ)/poly(ethylene oxide)(PEO)/Glutinous rice starch (GRS) nanofibers by the electrospinning method. A number of parameters such as the ratio between PEO and GRS and calcination temperature have been studied. The as-spun PZ/PEO/GRS composite and PZ fibers were characterized by TG-DTA, X-ray diffraction, FT-IR and SEM, respectively. SEM results showed that smooth and continuous fibers were obtained at the volume ratio of 10:2:1, PZ/PEO/GRS. After calcination of the as-spun PZ/PEO/GRS composite nanofibers at 650 degrees C for 4 h, PZ nanofibers with perovskite structure were obtained successfully. The fibers had a uniform and smooth surface without grain boundaries. However, when the calcination temperature increased to 750 degrees C and 850 degrees C, the fiber represented a necklace-like structure with grain boundaries arranged by grain to grain unit cell clusters.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble