Merck
  • Effect of alkalinisation of lignocaine for propofol injection pain: a prospective, randomised, double-blind study.
Effect of alkalinisation of lignocaine for propofol injection pain: a prospective, randomised, double-blind study.
Anaesthesia and intensive care (2013-07-03)
U Ozgul, Z Begec, M A Erdogan, M S Aydogan, M Sanli, C Colak
ABSTRACT
The aim of this study was to determine whether pretreatment with alkalinised lignocaine reduced the incidence and severity of pain during propofol injection. This prospective, randomised, double-blind study included 300 adult, American Society of Anesthesiologists physcial status I to II patients undergoing elective surgery. Patients were randomly allocated to one of three groups: Group L received 0.05 ml/kg of 1% lignocaine (5 ml normal saline + 5 ml 2% lignocaine), Group A received 0.05 ml/kg alkalinised lignocaine (5 ml 2% lignocaine + 1 ml 8.4% NaHCO3 + 4 ml normal saline), and Group S, the control group, was given the same amount of normal saline (NaCl 0.9%). All drugs were given as a bolus over 20 seconds before propofol administration. A blinded researcher assessed the patient's pain level using a four-point scale. The pain score [median (range)] and the incidence of pain in Group A (6%) was significantly lower than in groups L (41%) and S (88%, P <0.001). In addition, the pain score and the incidence of pain were found to be significantly different between Group L and Group S (P <0.001). The incidence of moderate and severe pain were greater in Group S when compared with groups A and L (P <0.001). Intravenous pretreatment with alkalinised lignocaine appears to be effective in reducing the pain during propofol injection.
Product Description
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C