Merck
  • ESA frequency and hemoglobin levels in patients on peritoneal dialysis: 2002 vs. 2008.
ESA frequency and hemoglobin levels in patients on peritoneal dialysis: 2002 vs. 2008.
Nephrology nursing journal : journal of the American Nephrology Nurses' Association (2013-06-19)
Ann Kruger, Lynette Trowbridge, Jane York, Belinda Butcher, Jennifer Bradley
ABSTRACT
This study examined whether a change infrequency of administration of erythropoietin-stimulating agent affected hemoglobin levels in patients on peritoneal dialysis. Data were extracted from the Australian Renal Anaemia Management database for the years 2002 and 2008. Less frequent dosing and increasing age were associated with higher hemoglobin levels, while increasing ferritin levels and later years were associated with lower hemoglobin levels.
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Pricing and availability is not currently available.
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Pricing and availability is not currently available.
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Pricing and availability is not currently available.