Merck
  • Selumetinib plus dacarbazine versus placebo plus dacarbazine as first-line treatment for BRAF-mutant metastatic melanoma: a phase 2 double-blind randomised study.
Selumetinib plus dacarbazine versus placebo plus dacarbazine as first-line treatment for BRAF-mutant metastatic melanoma: a phase 2 double-blind randomised study.
The Lancet. Oncology (2013-06-06)
Caroline Robert, Reinhard Dummer, Ralf Gutzmer, Paul Lorigan, Kevin B Kim, Marta Nyakas, Ana Arance, Gabriella Liszkay, Dirk Schadendorf, Mireille Cantarini, Stuart Spencer, Mark R Middleton
ABSTRACT
Patients with metastatic melanoma, 50% of whose tumours harbour a BRAF mutation, have a poor prognosis. Selumetinib, a MEK1/2 inhibitor, has shown antitumour activity in patients with BRAF-mutant melanoma and in preclinical models when combined with chemotherapy. This study was designed to look for a signal of improved efficacy by comparing the combination of selumetinib and dacarbazine with dacarbazine alone. This double-blind, randomised, placebo-controlled phase 2 study investigated selumetinib plus dacarbazine versus placebo plus dacarbazine as first-line treatment in patients older than 18 years with histologically or cytologically confirmed advanced BRAF-mutant cutaneous or unknown primary melanoma. Patients were randomly assigned by central interactive voice response system (1:1 ratio, block size four) to take either oral selumetinib (75 mg twice daily in a 21-day cycle) or placebo; all patients received intravenous dacarbazine (1000 mg/m(2) on day 1 of a 21-day cycle). Patients, investigators, and the study team were masked to the treatment assigned. The primary endpoint was overall survival analysed by intention to treat. This study is registered at ClinicalTrials.gov, NCT00936221. Between July 20, 2009, and April 8, 2010, 91 patients were randomly assigned to receive dacarbazine in combination with selumetinib (n=45) or placebo (n=46). Overall survival did not differ significantly between groups (median 13·9 months, 80% CI 10·2-15·6, in the selumetinib plus dacarbazine group and 10·5 months, 9·6-14·7, in the placebo plus dacarbazine group; hazard ratio [HR] 0·93, 80% CI 0·67-1·28, one-sided p=0·39). However, progression-free survival was significantly improved in the selumetinib plus dacarbazine group versus the placebo plus dacarbazine group (HR 0·63, 80% CI 0·47-0·84, one-sided p=0·021), with a median of 5·6 months (80% CI 4·9-5·9) versus 3·0 months (2·8-4·6), respectively. The most frequent adverse events included nausea (28 [64%] of 44 patients on selumetinib vs 25 [56%] of 45 on placebo), acneiform dermatitis (23 [52%] vs one [2%]), diarrhoea (21 [48%] vs 13 [29%]), vomiting (21 [48%] vs 15 [33%]), and peripheral oedema (19 [43%] vs three [7%]). The most common grade 3-4 adverse event was neutropenia (six [14%] patients in the selumetinib plus dacarbazine group vs four [9%] in the placebo plus dacarbazine group). Selumetinib plus dacarbazine showed clinical activity in patients with BRAF-mutant cutaneous or unknown primary melanoma, reflected by a significant benefit in progression-free survival compared with placebo plus dacarbazine group, although no significant change in overall survival was noted. The tolerability of this combination was generally consistent with monotherapy safety profiles. AstraZeneca.
Sigma-Aldrich
Dacarbazine, antineoplastic purine analog
Pricing and availability is not currently available.