Merck
Food microstructure and starch digestion.
Advances in food and nutrition research (2013-06-01)
Jaspreet Singh, Lovedeep Kaur, Harjinder Singh
ABSTRACT
Microstructural characteristics of starch-based natural foods such as parenchyma or cotyledon cell shape, cell size and composition, and cell wall composition play a key role in influencing the starch digestibility during gastrointestinal digestion. The stability of cell wall components and the arrangement of starch granules in the cells may affect the free access of amylolytic enzymes during digestion. Commonly used food processing techniques such as thermal processing, extrusion cooking, and post-cooking refrigerated storage alter the physical state of starch (gelatinization, retrogradation, etc.) and its digestibility. Rheological characteristics (viscosity) of food affect the water availability during starch hydrolysis and, consequently, the absorption of digested carbohydrates in the gastrointestinal tract. The nonstarch ingredients and other constituents present in food matrix, such as proteins and lipids interact with starch during processing, which leads to an alteration in the overall starch digestibility and physicochemical characteristics of digesta. Starch digestibility can be controlled by critically manipulating the food microstructure, processing techniques, and food composition.
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble