Merck
  • Targeted phosphotyrosine profiling of glycoprotein VI signaling implicates oligophrenin-1 in platelet filopodia formation.
Targeted phosphotyrosine profiling of glycoprotein VI signaling implicates oligophrenin-1 in platelet filopodia formation.
Arteriosclerosis, thrombosis, and vascular biology (2013-04-27)
Onno B Bleijerveld, Thijs C van Holten, Christian Preisinger, Jasper J van der Smagt, Richard W Farndale, Tjitske Kleefstra, Marjolein H Willemsen, Rolf T Urbanus, Philip G de Groot, Albert J R Heck, Mark Roest, Arjen Scholten
ABSTRACT
Platelet adhesion to subendothelial collagen is dependent on the integrin α2β1 and glycoprotein VI (GPVI) receptors. The major signaling routes in collagen-dependent platelet activation are outlined; however, crucial detailed knowledge of the actual phosphorylation events mediating them is still limited. Here, we explore phosphotyrosine signaling events downstream of GPVI with site-specific detail. Immunoprecipitations of phosphotyrosine-modified peptides from protein digests of GPVI-activated and resting human platelets were compared by stable isotope-based quantitative mass spectrometry. We surveyed 214 unique phosphotyrosine sites over 2 time points, of which 28 showed a significant increase in phosphorylation on GPVI activation. Among these was Tyr370 of oligophrenin-1 (OPHN1), a Rho GTPase-activating protein. To elucidate the function of OPHN1 in platelets, we performed an array of functional platelet analyses within a small cohort of patients with rare oligophrenia. Because of germline mutations in the OPHN1 gene locus, these patients lack OPHN1 expression entirely and are in essence a human knockout model. Our studies revealed that among other unaltered properties, patients with oligophrenia show normal P-selectin exposure and αIIbβ3 activation in response to GPVI, as well as normal aggregate formation on collagen under shear conditions. Finally, the major difference in OPHN1-deficient platelets turned out to be a significantly reduced collagen-induced filopodia formation. In-depth phosphotyrosine screening revealed many novel signaling recipients downstream of GPVI activation uncovering a new level of detail within this important pathway. To illustrate the strength of such data, functional follow-up of OPHN1 in human platelets deficient in this protein showed reduced filopodia formation on collagen, an important parameter of platelet hemostatic function.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)