Merck
  • [Simplified, low-cost gene expression profiling for the prediction of outcome in breast cancer based on routine histologic specimens].
[Simplified, low-cost gene expression profiling for the prediction of outcome in breast cancer based on routine histologic specimens].
Orvosi hetilap (2013-04-17)
A Marcell Szász, Balázs Ács, Emese Ágoston, Zsófia Sztupinszki, Anna-Mária Tőkés, Liliána Szittya, Borbála Székely, Miklós Szendrői, Qiyuan Li, László Harsányi, József Tímár, Zoltán Szállási, Charles Swanton, Balázs Győrffy, Janina Kulka
ABSTRACT
Grade 2 breast carcinomas do not form a uniform prognostic group. To extend the number of patients and the investigated genes of a previously identified prognostic signature described by the authors that reflect chromosomal instability in order to refine characterization of grade 2 breast cancers and identify driver genes. Using publicly available databases, the authors selected 9 target and 3 housekeeping genes that are capable to divide grade 2 breast carcinomas into prognostic groups. Gene expression was investigated by polymerase chain reaction in 249 formalin-fixed, paraffin-embedded breast tumors. The results were correlated with relapse-free survival. Histologically grade 2 carcinomas were split into good and a poor prognosis groups. Centroid-based ranking showed that 3 genes, FOXM1, TOP2A and CLDN4 were able to separate the good and poor prognostic groups of grade 2 breast carcinomas. Using appropriately selected control genes, a limited set of genes is able to split prognostic groups of breast carcinomas independently from their grade.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O